missing translation for 'onlineSavingsMsg'
Learn More

Afatinib dimaleate, MedChemExpress

Product Code. 16473748
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Product Code. Quantity unitSize
16473748 10 mg 10mg
16444816 100 mg 100mg
16410137 50 mg 50mg
16450966 200 mg 200mg
4 options
This item is not returnable. View return policy

Product Code. 16473748

Brand: Medchem Express HY10261A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Afatinib (BIBW 2992) dimaleate is an orally active, potent and irreversible dual specificity inhibitor of ErbB family (EGFR and HER2), with IC50 values of 0.5 nM, 0.4 nM, 10 nM and 14 nM for EGFRwt, EGFRL858R, EGFRL858R/T790M and HER2, respectively. Afatinib dimaleate can be used for the research of esophageal squamous cell carcinoma (ESCC), non-small cell lung cancer (NSCLC) and gastric cancer.

Afatinib (BIBW 2992) dimaleate is an orally active, potent and irreversible dual specificity inhibitor of ErbB family (EGFR and HER2), with IC50 values of 0.5 nM, 0.4 nM, 10 nM and 14 nM for EGFRwt, EGFRL858R, EGFRL858R/T790M and HER2, respectively. Afatinib dimaleate can be used for the research of esophageal squamous cell carcinoma (ESCC), non-small cell lung cancer (NSCLC) and gastric cancer.

Specifications

Chemical Name or Material Afatinib dimaleate
CAS 850140-73-7
Color White
Purity Grade Notes Research
Molecular Formula C32H33ClFN5O11
Quantity 10 mg
Synonym BIBW 2992MA2
Solubility Information H2O : 50 mg/mL (69.63 mM; Need ultrasonic) ∣DMSO : ≥ 35 mg/mL (48.74 mM)
SMILES O=C(NC1=C(C=C2C(C(NC3=CC(Cl)=C(C=C3)F)=NC=N2)=C1)O[C@H]4CCOC4)/C=C/CN(C)C.O=C(O)/C=C\C(O)=O.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 718.08
Formula Weight 718.08
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.