Learn More
Keystone Cleanroom Products 1073B Tyvek™ Autoclavable Stopper Bags
BIOPHARMAGREENER_CHOICE
Product Code. 17778497 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
12 x 12 in.
12 x 19 in.
19 x 20 in.
20 x 26 in.
Unit Size:
Pack of 500
Product Code. 17778497
Brand: Keystone Cleanroom Products SBMGTVKHS2026
Only null left
Add to basket
Add to basket
Keystone Cleanroom Products 1073B Tyvek™ Autoclavable Stopper Bags are breathable, and provide an effective bacterial barrier
  • Made from DuPont Tyvek and a polyethylene/HDPE film
  • This combination of materials yields superior seal strength for stoppers, seals, and other critical components, or equipment parts
  • These autoclavable bags are lint-free, moisture resistant, puncture resistant, and extremely tough
  • Tyvek™ side offers high breathability while providing an effective bacterial barrier
  • Clear film side offers excellent clarity combined with strength and durability
  • Can be used in gas or steam sterilizers; and can also be irradiated
  • Bags should be heat sealed closed, using a validate-able impulse, or continuous band sealer
Specifications
Description 1073B Tyvek™
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Heat Seal
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Quantity 500/Pk.
Dimensions (L x W) 20 x 26 in.
Length (English) 26 in.
Length (Metric) 66 cm
Width (English) 20 in.
Width (Metric) 50.8 cm
Product Type Autoclave Bag