Learn More
Keystone Cleanroom Products 1073B Tyvek™ Autoclavable Stopper Bags
Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
12 x 12 in.
12 x 19 in.
19 x 20 in.
20 x 26 in.
Unit Size:
Pack of 500
Description
- Made from DuPont Tyvek and a polyethylene/HDPE film
- This combination of materials yields superior seal strength for stoppers, seals, and other critical components, or equipment parts
- These autoclavable bags are lint-free, moisture resistant, puncture resistant, and extremely tough
- Tyvek™ side offers high breathability while providing an effective bacterial barrier
- Clear film side offers excellent clarity combined with strength and durability
- Can be used in gas or steam sterilizers; and can also be irradiated
- Bags should be heat sealed closed, using a validate-able impulse, or continuous band sealer
Specifications
Specifications
| Description | 1073B Tyvek™ |
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance (COC) |
| Closure Type | Heat Seal |
| Color | White |
| Disposable | Single-use |
| For Use With (Application) | Steam Sterilization |
| Material | Tyvek |
| Sterility | Non-sterile |
| Latex Free | Yes |
| Show More |