Learn More
Kimberly-Clark™ Kimtech™ WetTask™ SXX Wipers
Product Code. 12605636 Shop All Kimberly-Clark Professional Products
Click to view available options
Dimensions (L x W):
15.2 x 22.5 cm
Unit Size:
6 rolls
Product Code. 12605636
Brand: Kimberly-Clark™ 7769
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Kimtech™ WetTask SXX Wipers are ideal for solvent use, surface preparation and wiping harsh chemicals in industrial and healthcare sectors.
  • 6 x 175 white sheets (1,050 total)
  • Low-lint
  • Silicone free
  • pH stable
  • Ideal for solvent use, surface preparation and wiping harsh chemicals
  • Designed with the aviation transport sector
  • Certified to requirements of BMS 15-5F and AMZ 3819B
TRUSTED_SUSTAINABILITY
Specifications
Product Type Light-duty Industrial Wiper
Length (English) 6 in.
Length (Metric) 15.2 cm
Width (English) 8.9 in.
Width (Metric) 22.5 cm
Color White
Dimensions (L x W) 15.2 x 22.5 cm
For Use With (Equipment) WetTask™ Roll Wiper Dispenser