missing translation for 'onlineSavingsMsg'
Learn More

ZCL278, MedChemExpress

Product Code. 16416376
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16416376

Brand: Medchem Express HY13963

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZCL278 is a selective Cdc42 modulator that directly binds to Cdc42 and inhibits its functions with Kd of 11.4 μM for Cdc42-ZCL278 affinity in surface plasmon resonance (SPR) experiment.

ZCL278 is a selective Cdc42 modulator that directly binds to Cdc42 and inhibits its functions with Kd of 11.4 μM for Cdc42-ZCL278 affinity in surface plasmon resonance (SPR) experiment.

Specifications

Chemical Name or Material ZCL278
CAS 587841-73-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H19BrClN5O4S2
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (85.49 mM; Need ultrasonic)
SMILES O=C(NC(NC1=CC=C(S(=O)(NC2=NC(C)=CC(C)=N2)=O)C=C1)=S)COC3=CC=C(Br)C=C3Cl
Molecular Weight (g/mol) 584.89
Formula Weight 584.89
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.