missing translation for 'onlineSavingsMsg'
Learn More

Teprenone, MedChemExpress

Product Code. 16438456
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438456 100 mg 100mg
16412216 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438456 Supplier Medchem Express Supplier No. HYB0779

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Teprenone is an anti-ulcer drug, and works as an inducer of heat shock proteins (HSPs).

Teprenone is an anti-ulcer drug, and works as an inducer of heat shock proteins (HSPs).

Specifications

Chemical Name or Material Teprenone
Physical Form Liquid
CAS 6809-52-5
Color Colorless
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C23H38O
For Use With (Application) Cancer-programmed cell death
Synonym Geranylgeranylacetone
Solubility Information DMSO : 100 mg/mL (302.53 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (151.26 mM; Need ultrasonic)
SMILES CC(CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C)=O
Molecular Weight (g/mol) 330.55
Formula Weight 330.55
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.