missing translation for 'onlineSavingsMsg'
Learn More

Quinacrine dihydrochloride, MedChemExpress

Product Code. 16496296
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
16496296 100 mg 100mg
16481696 500 mg 500mg
2 options
This item is not returnable. View return policy

Product Code. 16496296

Brand: Medchem Express HY13735A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Quinacrine (Mepacrine) dihydrochloride is an orally bioavailable antimalarial agent, which possess anticancer effect both in vitro and vivo. Quinacrine dihydrochloride suppresses NF-κB and activate p53 signaling, which results in the induction of the apoptosis.

Quinacrine (Mepacrine) dihydrochloride is an orally bioavailable antimalarial agent, which possess anticancer effect both in vitro and vivo. Quinacrine dihydrochloride suppresses NF-κB and activate p53 signaling, which results in the induction of the apoptosis.

Specifications

Chemical Name or Material Quinacrine dihydrochloride
Physical Form Solid
CAS 69-05-6
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C23H32Cl3N3O
For Use With (Application) Cancer-programmed cell death
Synonym Mepacrine dihydrochlorideSN-390 dihydrochloride
Solubility Information DMSO : ≥ 44 mg/mL (93.05 mM) ∣H2O : 13.89 mg/mL (29.37 mM; Need ultrasonic)
SMILES CC(NC1=C(C=C(OC)C=C2)C2=NC3=CC(Cl)=CC=C31)CCCN(CC)CC.Cl.Cl
Molecular Weight (g/mol) 472.88
Formula Weight 472.88
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.