missing translation for 'onlineSavingsMsg'
Learn More

PF 05089771, MedChemExpress

Product Code. 16476026
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
100 mg
Unit Size:
100mg
5mg
Product Code. Quantity unitSize
16476026 100 mg 100mg
16458147 5 mg 5mg
2 options
This item is not returnable. View return policy

Product Code. 16476026

Brand: Medchem Express HY12883

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF 05089771 is a potent, orally active and selective arylsulfonamide Nav1.7 inhibitor, with IC50 values of 11 nM, 12 nM, 13 nM, 171 nM and 8 nM for hNav1.7, cynNav1.7, dogNav1.7, ratNav1.7, and musNav1.7, respectively. PF 05089771 is under the study for pain and diabetic neuropathy.

PF 05089771 is a potent, orally active and selective arylsulfonamide Nav1.7 inhibitor, with IC50 values of 11 nM, 12 nM, 13 nM, 171 nM and 8 nM for hNav1.7, cynNav1.7, dogNav1.7,

Specifications

Chemical Name or Material PF 05089771
CAS 1235403-62-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H12Cl2FN5O3S2
Quantity 100 mg
Solubility Information DMSO : ≥ 34 mg/mL (67.95 mM)
SMILES O=S(C1=CC(Cl)=C(OC2=CC=C(Cl)C=C2C3=CNN=C3N)C=C1F)(NC4=CSC=N4)=O
Molecular Weight (g/mol) 500.35
Formula Weight 500.35
Percent Purity 98.86%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.