missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olanzapine , MedChemExpress
Description
Olanzapine (LY170053) is a selective, orally active monoaminergic antagonist with high affinity binding to serotonin H1, 5HT2A/2C, 5HT3, 5HT6 (Ki=7, 4, 11, 57, and 5 nM, respectively), dopamine D1-4 (Ki=11 to 31 nM), muscarinic M1-5 (Ki=1.9-25 nM), and adrenergic α1 receptor (Ki=19 nM). Olanzapine is an atypical antipsychotic.
Specifications
Specifications
| Chemical Name or Material | Olanzapine |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H20N4S |
| Quantity | 100 mg |
| Formula Weight | 312.43 |
| Grade | Research |
| Physical Form | Solid |
| For Use With (Application) | Cancer-programmed cell death |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?