missing translation for 'onlineSavingsMsg'
Learn More

OICR-9429, MedChemExpress

Product Code. 16447216
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16447216 100 mg 100mg
16457248 10 mg 10mg
16429777 5 mg 5mg
16423566 25 mg 25mg
16423267 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16447216

Brand: Medchem Express HY16993

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OICR-9429 is high affinity WD repeat domain 5 (WDR5) inhibitor, competitively blocks WDR5 interaction with MLL protein via binding the central peptide-binding pocket of WDR5. OICR-9429 can suppress histone H3K4 trimethylation and can be used for the research of various cancers including non-MLL-rearranged leukaemia, colon, pancreatic, prostate cancer and bladder cancer (BCa) .

OICR-9429 is high affinity WD repeat domain 5 (WDR5) inhibitor, competitively blocks WDR5 interaction with MLL protein via binding the central peptide-binding pocket of WDR5. OICR-9429 can suppress histone H3K4 trimethylation and can be used for the research of various cancers including non-MLL-rearranged leukaemia, colon, pancreatic, prostate cancer and bladder cancer (BCa) .

Specifications

Chemical Name or Material OICR-9429
Physical Form Solid
CAS 1801787-56-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C29H32F3N5O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 32 mg/mL (57.60 mM)
SMILES O=C(C1=CNC(C=C1C(F)(F)F)=O)NC2=CC(C3=CC(CN4CCOCC4)=CC=C3)=CC=C2N5CCN(C)CC5
Molecular Weight (g/mol) 555.59
Formula Weight 555.59
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.