missing translation for 'onlineSavingsMsg'
Learn More

Ningetinib Tosylate, MedChemExpress

Product Code. 16435076
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16435076 100 mg 100mg
16490457 50 mg 50mg
16456807 5 mg 5mg
16414108 10 mg 10mg
16402956 25 mg 25mg
5 options
This item is not returnable. View return policy

Product Code. 16435076

Brand: Medchem Express HY107145

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ningetinib Tosylate is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and c-Met, VEGFR2 and Axl, respectively.

Ningetinib Tosylate is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and c-Met, VEGFR2 and Axl, respectively.

Specifications

Chemical Name or Material Ningetinib Tosylate
CAS 1394820-77-9
Color White
Purity Grade Notes Research
Molecular Formula C38H37FN4O8S
Quantity 100 mg
Solubility Information DMSO : 33.33 mg/mL (45.73 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC=C(S(=O)(O)=O)C=C1.CC2=C(C(NC3=CC(F)=C(OC4=CC=NC5=C4C=CC(OCC(C)(O)C)=C5)C=C3)=O)C(N(C6=CC=CC=C6)N2C)=O
Molecular Weight (g/mol) 728.79
Formula Weight 728.79
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.