missing translation for 'onlineSavingsMsg'
Learn More

Mycro 3, MedChemExpress

Product Code. 16484406
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484406

Brand: Medchem Express HY100669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mycro 3 is a potent and selective inhibitor of Myc-associated factor X (MAX) dimerization. Mycro 3 also inhibit DNA binding of c-Myc. Mycro 3 could be used for the research of pancreatic cancer.

Mycro 3 is a potent and selective inhibitor of Myc-associated factor X (MAX) dimerization. Mycro 3 also inhibit DNA binding of c-Myc. Mycro 3 could be used for the research of pancreatic cancer.

Specifications

Chemical Name or Material Mycro 3
Physical Form Solid
CAS 944547-46-0
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C24H17ClF2N6O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (189.80 mM)
SMILES O=C(C1=C(NC(C2=NN3C(C(F)(Cl)F)=CC(C4=CC=CO4)=NC3=C2)=O)N(C5=CC=CC=C5)N=C1)OCC
Molecular Weight (g/mol) 526.88
Formula Weight 526.88
Percent Purity 99.21%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.