missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ML-030, MedChemExpress
Description
ML-030 is a potent PDE4 inhibitor, with IC50 of 6.7 nM, 12.9 nM, 48.2 nM, 37.2 nM, 452 nM and 49.2 nM for PDE4A, PDE4A1, PDE4B1, PDE4B2, PDE4C1,and PDE4D2, respectively.
Specifications
Specifications
| Chemical Name or Material | ML-030 |
| CAS | 1013750-77-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H20N4O4S |
| Quantity | 100 mg |
| Solubility Information | DMSO : 50 mg/mL (121.22 mM; Need ultrasonic) |
| SMILES | COC1=CC=C(C2=NN3C(SC2)=NN=C3C4=CC(OC)=CC=C4OC)C=C1OC |
| Molecular Weight (g/mol) | 412.46 |
| Formula Weight | 412.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?