Learn More
Minocycline hydrochloride, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Minocycline hydrochloride is a hypoxia-inducible factor (HIF)-1α inhibitor. Minocycline hydrochloride shows anti-cancer, anti-inflammatory, and glutamate antagonist effects. Minocycline hydrochloride reduces glutamate neurotransmission and shows neuroprotective properties and antidepressant effects. Minocycline hydrochloride inhibits bacterial protein synthesis through binding with the 30S subunit of the bacterial ribosome, resulting in a bacteriostatic effect.
Specifications
Specifications
| Chemical Name or Material | Minocycline hydrochloride |
| CAS | 13614-98-7 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H28ClN3O7 |
| Quantity | 100 mg |
| Solubility Information | H2O : 10 mg/mL (20.25 mM; Need ultrasonic) ∣DMSO : 0.49 mg/mL (0.99 mM; Need ultrasonic and warming) |
| SMILES | OC1=C(C(N)=O)C([C@@]2(O)C(O)=C3C(C4=C(O)C=CC(N(C)C)=C4C[C@@]3([H])C[C@@]2([H])[C@@H]1N(C)C)=O)=O.Cl |
| Molecular Weight (g/mol) | 493.94 |
| Formula Weight | 493.94 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.