missing translation for 'onlineSavingsMsg'
Learn More

Magnolol, MedChemExpress

Product Code. 16419016
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16419016

Brand: Medchem Express HYN0163

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Magnolol, a natural lignan isolated from the stem bark of Magnolia officinalis, is a dual agonist of both RXRα and PPARγ, with EC50 values of 10.4 μM and 17.7 μM, respectively.

Magnolol, a natural lignan isolated from the stem bark of Magnolia officinalis, is a dual agonist of both RXRα and PPARγ, with EC50 values of 10.4 μM and 17.7 μM, respectively.

Specifications

Chemical Name or Material Magnolol
CAS 528-43-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H18O2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (375.47 mM; Need ultrasonic)
SMILES OC1=CC=C(CC=C)C=C1C2=CC(CC=C)=CC=C2O
Molecular Weight (g/mol) 266.33
Formula Weight 266.33
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.