missing translation for 'onlineSavingsMsg'
Learn More

LY2940094, MedChemExpress

Product Code. 16475626
Encompass
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475626

Brand: Medchem Express HY114452

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress LY2940094 (BTRX-246040) is a potent, selective and orally available nociceptin receptor (NOP receptor) antagonist with high affinity (Ki=0.105 nM) and antagonist potency (Kb=0.166 nM). LY2940094 reduces ethanol self-administration in animal models.

LY2940094 (BTRX-246040) is a potent, selective and orally available nociceptin receptor (NOP receptor) antagonist with high affinity (Ki=0.105 nM) and antagonist potency (Kb=0.166 nM). LY2940094 reduces ethanol self-administration in animal models.

Specifications

Chemical Name or Material LY2940094
CAS 1307245-86-8
Color White
Purity Grade Notes Research
Molecular Formula C22H23ClF2N4O2S
Quantity 100 mg
Synonym BTRX-246040
Solubility Information DMSO : 41.67 mg/mL (86.64 mM; Need ultrasonic)
SMILES OCC1=CC=CN=C1N2N=C(C)C(CN3CCC4(C5=C(C=C(Cl)S5)C(F)(F)CO4)CC3)=C2
Molecular Weight (g/mol) 480.96
Formula Weight 480.96
Percent Purity 98.1%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.