missing translation for 'onlineSavingsMsg'
Learn More

Lornoxicam, MedChemExpress

Product Code. 16498256
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16498256

Brand: Medchem Express HYB0367

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lornoxicam (Chlortenoxicam), a COX-1 and COX-2 inhibitor, is a new nonsteroidal anti-inflammatory drug (NSAID).

Lornoxicam (Chlortenoxicam), a COX-1 and COX-2 inhibitor, is a new nonsteroidal anti-inflammatory drug (NSAID).

Specifications

Chemical Name or Material Lornoxicam
CAS 70374-39-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H10ClN3O4S2
Quantity 100 mg
Synonym Chlortenoxicam Ro 13-9297
Solubility Information DMSO : 3.8 mg/mL (10.22 mM; Need ultrasonic and warming)
SMILES O=C(C1=C(O)C2=C(C=C(Cl)S2)S(N1C)(=O)=O)NC3=NC=CC=C3
Molecular Weight (g/mol) 371.82
Formula Weight 371.82
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.