missing translation for 'onlineSavingsMsg'
Learn More

Ilorasertib, MedChemExpress

Product Code. 16427046
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. Quantity unitSize
16427046 100 mg 100mg
16433027 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 16427046

Brand: Medchem Express HY16018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ilorasertib (ABT-348) is a potent and ATP-competitive multitargeted kinase inhibitor, which inhibits Aurora C, Aurora B, and Aurora A with IC50s of 1 nM, 7 nM, 120 nM, respectively. Ilorasertib also suppresses RET tyrosine kinase, PDGFRβ and Flt1 with IC50s of 7 nM, 3 nM and 32 nM, respectively.

Ilorasertib (ABT-348) is a potent and ATP-competitive multitargeted kinase inhibitor, which inhibits Aurora C, Aurora B, and Aurora A with IC50s of 1 nM, 7 nM, 120 nM, respectively. Ilorasertib also suppresses RET tyrosine kinase, PDGFRβ and Flt1 with IC50s of 7 nM, 3 nM and 32 nM, respectively.

Specifications

Chemical Name or Material Ilorasertib
CAS 1227939-82-3
Color Brown
Purity Grade Notes Research
Molecular Formula C25H21FN6O2S
Quantity 100 mg
Synonym ABT-348
Solubility Information DMSO : 41.67 mg/mL (85.29 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CC(F)=C1)NC2=CC=C(C3=CSC4=C3C(N)=NC=C4C5=CN(CCO)N=C5)C=C2
Molecular Weight (g/mol) 488.54
Formula Weight 488.54
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.