missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Halobetasol (propionate), MedChemExpress
Description
Halobetasol propionate is a synthetic corticosteroid for topical dermatological use
Specifications
Specifications
| Chemical Name or Material | Halobetasol (propionate) |
| CAS | 66852-54-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H31ClF2O5 |
| Quantity | 100 mg |
| Synonym | BMY-30056 CGP-14458 Ulobetasol propionate |
| Solubility Information | DMSO : ≥ 100 mg/mL (206.20 mM) |
| SMILES | C[C@@]12[C@](C(CCl)=O)(OC(CC)=O)[C@@H](C)C[C@@]1([H])[C@]3([H])C[C@H](F)C4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C2)=O |
| Molecular Weight (g/mol) | 484.96 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?