missing translation for 'onlineSavingsMsg'
Learn More

Gadopentetate dimeglumine, MedChemExpress

Product Code. 16498036
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Product Code. Quantity unitSize
16498036 100 mg 100mg
16461356 200 mg 200mg
16441986 500 mg 500mg
16434197 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16498036

Brand: Medchem Express HYA0167

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Gadopentetate dimeglumine (SH-L-451A) is used in combination with magnetic resonance imaging (MRI) to allow blood vessels, organs, and other non-bony tissues to be seen more clearly on the MRI.

Gadopentetate dimeglumine (SH-L-451A) is used in combination with magnetic resonance imaging (MRI) to allow blood vessels, organs, and other non-bony tissues to be seen more clearly on the MRI.

Specifications

Chemical Name or Material Gadopentetate dimeglumine
Physical Form Solid
CAS 86050-77-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C28H54GdN5O20
For Use With (Application) Cancer-programmed cell death
Synonym SH-L-451A Gadopentetic acid dimeglumine salt
Solubility Information DMSO : 100 mg/mL (106.61 mM; Need ultrasonic)
SMILES O=C1[O-][Gd+3]([N]2(C3)CC4=O)([N]5(C6)CC7=O)([O-]C6=O)([O-]7)([O-]C3=O)([O-]4)[N](CC5)(CC2)C1.O[C@H]([C@@H](O)CNC)[C@H](O)[C@H](O)CO.O[C@H]([C@@H](O)CNC)[C@H](O)[C@H](O)CO.[2H+]
Molecular Weight (g/mol) 938.0
Formula Weight 938.0
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.