missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxylamine succinate, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Doxylamine (succinate), a first generation antihistamine, is a histamine (H1) receptor antagonist. Doxylamine is also a local analgesic agent and effective hypnotic agent.
Specifications
Specifications
| Chemical Name or Material | Doxylamine succinate |
| Physical Form | Solid |
| CAS | 562-10-7 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 100 mg/mL (257.43 mM) ∣H2O : 50 mg/mL (128.71 mM; Need ultrasonic) |
| SMILES | O=C(O)CCC(O)=O.CC(C1=NC=CC=C1)(C2=CC=CC=C2)OCCN(C)C |
| Molecular Weight (g/mol) | 388.46 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction