Learn More
(3Alpha,5Beta,7Beta)-Cholane-3,7,24-triol, TRC
Product Code. 30008863
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008863 100 mg 100mg
30007864 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30008863 Supplier Toronto Research Chemicals Supplier No. TRCC292530100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 130593-75-8
Molecular Formula C24 H42 O3
Molecular Weight (g/mol) 378.59
Synonym 5β-Cholane-3α,7β,24-triol, (3α,5β,7β)-Cholane-3,7,24-triol
IUPAC Name (3R,5S,7S,8R,9S,10S,13R,14S,17R)-17-[(2R)-5-hydroxypentan-2-yl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,7-diol
SMILES C[C@H](CCCO)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C24H42O3/c1-15(5-4-12-25)18-6-7-19-22-20(9-11-24(18,19)3)23(2)10-8-17(26)13-16(23)14-21(22)27/h15-22,25-27H,4-14H2,1-3H3/t15-,16+,17-,18-,19+,20+,21+,22+,23+,24-/m1/s1
Formula Weight 378.3134
Percent Purity >95%
Chemical Name or Material (3alpha,5beta,7beta)-Cholane-3,7,24-triol
Grade HPLC