Learn More
KGW Isotherm™ Dewar Cylindrical Insulated Flasks
Description
- Easy handling
- Cooling of small samples
- Fully silvered as standard
- Flexible applications in laboratory
- Available with or without side grip
Specifications
Specifications
| Color | Blue |
| Diameter (Metric) Outer | 115mm |
| For Use With (Application) | Cooling of small samples, storage of liquid nitrogen, dry ice, other coolants and as insulating flask |
| Height (Metric) Inner | 245mm |
| Height (Metric) | 300mm |
| Includes | With side grip |
| Capacity (Metric) | 1500 mL |
| Diameter (Metric) Inner | 90 mm |
| Material | Borosilicate Glass |