missing translation for 'onlineSavingsMsg'
Learn More

1000 μg/g Ethylene Glycol Standard in Multi-grade Diesel Engine Oil, VHG™

Product Code. 17969124
Change view
Click to view available options
Quantity:
80 mL
Unit Size:
80mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17969124 80 mL 80mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17969124 Supplier VHG Labs Supplier No. VHGEGOIL100080
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

1000 μg/g Ethylene Glycol Standard in Multi-grade Diesel Engine Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylene glycol
CAS 107-21-1
Analyte or Component Names Ethylene glycol
Shipping Condition Room Temperature
Molecular Formula C2 H6 O2
Concentration 1000 μg/g
Synonym Hydroxyethyl Salicylate Imp. B (EP), Ethylene Glycol, Propan-1,2-diol, Glycerol Impurity B, Hydroxyethyl Salicylate Impurity B
InChI Formula InChI=1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
SMILES OCCO
IUPAC Name ethane-1,2-diol
Solvent or Matrix Multi-grade Diesel Engine Oil
Molecular Weight (g/mol) 62.07
Quantity 80 mL
Formula Weight 62.0368 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.