missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fibroblast Activation Protein alpha/FAP Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB9727N-100UG
This item is not returnable.
View return policy
Description
Fibroblast Activation Protein alpha/FAP Monoclonal antibody specifically detects Fibroblast Activation Protein alpha/FAP in Mouse samples. It is validated for Flow Cytometry
Specifications
| Fibroblast Activation Protein alpha/FAP | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Flow Cytometry | |
| 983802 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P97321 | |
| Rat | |
| RUO | |
| 2191 | |
| Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction