missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies 100 mm Tissue Culture-Treated Dishes
Click to view available options
Quantity:
10 Dishes
240 Dishes
Unit Size:
Pack of 10
Pack of 240
Description
- Tissue culture dishes, 100 mm, intended for culturing anchorage-dependent cells, including stromal cells
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Quantity | 10 Dishes |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction