Learn More
Hamilton™ 100 μL, Model 1710 TLL SYR, Long Life Instrument Syringe
Description
Specification Sheet
- Volume – 100 μL
- Syringe Type – Longlife Gastight
- Syringe Series – 1700
- Termination – PTFE Luer Lock (TLL)
- Plunger Style – X-Style
- Fluid Path – Borosilicate glass, PTFE, UHMWPE
- Autoclavable – No
- Gas Sterilizable – Yes
- Pressure – 1000 psig (68.9 bar)
- Maximum Temperature – 115°C / 239°F
- Minimum Temperature – 10°C / 50°F
- Barrel Inner Diameter – 0.057 inches (1.46 mm)
- Barrel Outer Diameter – 0.305 inches (7.75 mm)
- Std Needle Dead Volume – N/A
- Sales Quantity – 1 syringe per pack
Specifications
Specifications
| Product Type | Long Life UHMWPE Syringes, PSD/6 Syringes, Instrument Syringes |
| Volume (Metric) | 100 μL |