Learn More
10-Undecen-1-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 10-Undecen-1-ol |
| CAS | 112-43-6 |
| Molecular Formula | C11H22O |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C11H22O/c1-2-3-4-5-6-7-8-9-10-11-12/h2,12H,1,3-11H2 |
| SMILES | OCCCCCCCCCC=C |
| IUPAC Name | undec-10-en-1-ol |
| Molecular Weight (g/mol) | 170.29 |
| Formula Weight | 170.1671 |