Learn More
Hamilton™ 10 mL, Model 1010 TLL SYR, Long Life Instrument Syringe
Product Code. 12317479
Click to view available options
Volume (Metric):
10 mL
Unit Size:
Each
Product Code. 12317479
Brand: Hamilton™ 988250
The BFP syringe manufactured by Hamilton Company is intended for scientific research and laboratory use only and is not intended for human in vivo use.
Specification Sheet
  • Volume – 10 mL
  • Syringe Type – Longlife Gastight
  • Syringe Series – 1000
  • Termination – PTFE Luer Lock (TLL)
  • Plunger Style – Side Post Mounting (XL)
  • Fluid Path – Borosilicate glass, PTFE, UHMWPE
  • Autoclavable – No
  • Gas Sterilizable – Yes
  • Maximum Temperature – 115°C / 239°F
  • Minimum Temperature – 10°C / 50°F
  • Barrel Inner Diameter – 0.574 inches (14.6 mm)
  • Barrel Outer Diameter – 0.695 inches (17.7 mm)
  • Maximum Pressure – 200 psig (13.8 bar)
  • Sales Quantity – 1 syringe per pack
Specifications
Product Type Long Life UHMWPE Syringes, 1000 Series
Volume (Metric) 10 mL