Learn More
Haldenwanger X10 Melting Crucible DIN 12904 Medium 4 90ml Porcelain
Brand:  Haldenwanger 79MF/4
X10 Melting Crucible DIN 12904 Medium 4 90ml Porcelain
Product Code. 30215943
  • ONLINE EXCLUSIVE PRICE
    1415.00 NOK
    1187.86 NOK / Pack of 10
    Save 227.14 NOK
    16% Off
Only null left
Estimated Shipment: 02-02-2026