Learn More
Wheaton™ 10 μL Inoculation Loop
Description
- For dispersing samples on agar surface
- Meet quantitative standards for management of microbiological samples
Specifications
Specifications
| Disposable | Single-use |
| Volume | 10 μL |
| Material | ABS |
| Sterility | Sterile |
| Size | Standard |
| Color | Blue |
| Quantity | 500 |
| Packaging Type | Packaged in peel pouch |
| Autoclavable | Not Autoclavable |