Learn More
Hamilton™ 10 μL 1700 Series Gas Tight Syringe
Click to view available options
Needle Gauge:
22s G
23s G
26S (No Needle Included)
26s G
Custom
No Needle Included
Needle Point Style:
Blunt
Cone
Curved, Beveled (No Needle Included)
Custom
No Needle Included
Sharp, Bevel, Curved, Non-coring
Connection Type:
Cemented
Removable
Unit Size:
Each
Description
- This series of syringes are ideal for dispensing volumes up to 10 μL.
- A variety of different terminations and needle options are available in this series.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Needle Gauge | 22s G |
| Diameter (English) Inner | 0.018 in. |
| Diameter (English) Inner Needle | 0.006 in. |
| Diameter (Metric) Inner Needle | 0.15 mm |
| Diameter (English) Outer Needle | 0.0283 in. |
| Diameter (English) Outer | 0.260 in. |
| Diameter (Metric) Inner | 0.460 mm |
| Diameter (Metric) Outer | 6.60 mm |
| Show More |