Learn More
Vileda Professional™ 10 in. Aluminum Extendable Roll-O-Matic™ Mop w/attached Refill
Description
- Color coded tabs incorporated into the top of the handle allow for color coding of cleaning equipment
- Ergonomic grip reduces work fatigue
- Handle extends easily with turn of wrist from 58.5 to 76.25 in.
- Entire mop handle is completely autoclavable at the most common autoclave cycles
- Straight handle allows user to properly use the 'pull and lift' technique with both sides of the mop
- Perfect for use on floors, walls and ceilings
- Also available in 14 in.
Specifications
Specifications
| Product Type | Cleanroom Mop |
| Color | Blue, Silver |
| Length (English) | 10 in. |
| Autoclavable | Autoclavable |
| Quantity | 1/Cs. |
| For Use With (Equipment) | Any 10 in. Roll-O-Matic Sponge Refill |
| For Use With (Application) | Cleaning and Disinfecting Floors, Walls and Ceilings |
| Material | Galvanized Steel |