Learn More
1-Triethylsilyl-1-heptyne, 95%, Thermo Scientific™
Product Code. 15454437
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15454437 5 g 5g
15444437 1 g 1g
2 options
Product Code. 15454437
Brand: Thermo Scientific Alfa Aesar H31802.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1047660-72-9
Molecular Formula C13H26Si
Molecular Weight (g/mol) 210.436
MDL Number MFCD18800797
InChI Key KIKRQMVMQSYDCD-UHFFFAOYSA-N
Synonym 1-triethylsilyl-1-heptyne, triethyl hept-1-yn-1-yl silane
PubChem CID 99738219
IUPAC Name triethyl(hept-1-ynyl)silane
SMILES CCCCCC#C[Si](CC)(CC)CC
Specifications
Flash Point >85°C (185°F)
Linear Formula CH3(CH2)4C≡CSi(CH2CH3)3
Quantity 5 g
Formula Weight 210.43
Percent Purity 95%
Chemical Name or Material 1-Triethylsilyl-1-heptyne