Learn More
1-Tetradecanol, Sigma-Aldrich™
Product Code. 15879540
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879540 1 kg 1kg
15869540 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15879540 Supplier Sigma Aldrich Supplier No. 8081461000
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 112-72-1
Grade Research
Molecular Weight (g/mol) 214.39
Specifications
Melting Point 36°C to 39°C
Density 0.8 g/cm3 at 60°C
Boiling Point 277°C to 288°C/1013 hPa
MDL Number MFCD00004757
Quantity 1 kg
Synonym 1-Tetradecanol, Myristyl alcohol, Tetradecyl alcohol, 1-Tetradecanol