Learn More
1-Propanethiol, Sigma-Aldrich™
Product Code. 15884211
Change view
Click to view available options
Quantity:
5 mL
250 mL
Unit Size:
250mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884211 250 mL 250mL
15874211 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15884211 Supplier Sigma Aldrich Supplier No. 8075290250
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107-03-9
Grade Research
Molecular Weight (g/mol) 76.16
Specifications
Melting Point -113°C
Density 0.84 g/cm3 at 20°C
Boiling Point 67°C to 68°C/1013 hPa
MDL Number MFCD00004900
Quantity 250 mL
Synonym 1-Propanethiol, Propyl mercaptan, 1-Propanethiol