missing translation for 'onlineSavingsMsg'
Learn More

1-Propanesulfonyl chloride, 97%

Product Code. 15407248
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407248 100 mL 100mL
15497238 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407248 Supplier Thermo Scientific Alfa Aesar Supplier No. H60747.AE
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 10147-36-1 | C3H7ClO2S | 142.597 g/mol

1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Propanesulfonyl chloride was used in synthesis of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids.

Solubility
Decomposes in hot water.

Notes
Moisture sensitive. Store in dry temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10147-36-1
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.597
MDL Number MFCD00007462
InChI Key KPBSJEBFALFJTO-UHFFFAOYSA-N
Synonym 1-propanesulfonyl chloride, propanesulfonyl chloride, propanesulphonyl chloride, propylsulfonyl chloride, 1-propanesulfonylchloride, n-propanesulfonyl chloride, 1-propane sulfonyl chloride, chloropropylsulfone, n-prso2cl, propanesulfonylchloride
PubChem CID 66279
IUPAC Name propane-1-sulfonyl chloride
SMILES CCCS(=O)(=O)Cl

Specifications

Density 1.267
Boiling Point 78°C to 79°C
Flash Point 97°C (207°F)
Odor Pungent, Makes Eyes Water
Linear Formula CH3CH2CH2SO2Cl
Refractive Index 1.453
Quantity 100 mL
UN Number UN3265
Beilstein 1747500
Sensitivity Moisture sensitive
Solubility Information Decomposes in hot water.
Formula Weight 142.6
Percent Purity 97%
Chemical Name or Material 1-Propanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.