Learn More
1-Phenyl-1-pentanol, 95%
Product Code. 11331647
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11331647 10 g 10g
11341647 50 g 50g
2 options
Product Code. 11331647
Brand: Thermo Scientific Alfa Aesar L04646.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 583-03-9 | C11H16O | 164.248 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 583-03-9
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00021935
InChI Key OVGORFFCBUIFIA-UHFFFAOYSA-N
Synonym fenipentol, 1-phenyl-1-pentanol, 1-phenylpentanol, pancoral, phenylbutylcarbinol, suiclisin, alpha-butylbenzyl alcohol, 1-pentanol, 1-phenyl, 1-phenyl-1-hydroxypentane, ph bc
PubChem CID 3338
IUPAC Name 1-phenylpentan-1-ol
SMILES CCCCC(C1=CC=CC=C1)O
Specifications
Density 0.96
Boiling Point 137°C (2 mmHg)
Refractive Index 1.508
Quantity 10 g
Beilstein 1908761
Merck Index 14,3975
Formula Weight 164.25
Percent Purity 95%
Chemical Name or Material 1-Phenyl-1-pentanol