Learn More
1-Pentanesulfonic acid sodium salt, monohydrate; 99.9%, For HPLC analysis, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Specifications
Specifications
| Item Name | 1-Pentanesulfonic Acid, Sodium Salt, Monohydrate |
| Content And Storage | Store at Room Temperature. |
| Form | Crystalline Powder |
| Quantity | 100 g |
| Appearance | White |
| Molecular Formula | C5H11NaO3S |
| InChI Key | ROBLTDOHDSGGDT-UHFFFAOYSA-M |
| SMILES | [Na+].CCCCCS([O-])(=O)=O |
| IUPAC Name | sodium pentane-1-sulfonate |
| Molecular Weight (g/mol) | 174.19 |