missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanol, ACS reagent, ≥99%, Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2 L
2.5 L
4 L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | -15°C |
| Density | 0.827g/mL at 25°C |
| Boiling Point | 194°C |
| Flash Point | 80°C (176°F) |
| Odor | Characteristic |
| Packaging | Glass Bottle |
| Linear Formula | CH3(CH2)7OH |
| Refractive Index | n20/D 1.429 |
| Quantity | 2 L |
| Beilstein | 1697461 |
| Show More |
Safety and Handling
Product Identifier
- 1-Octanol
- Warning
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction