missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, ACS reagent, ≥99%, Honeywell™ Riedel-de Haën™

Product Code. 15615640
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2 L
2.5 L
4 L
This item is not returnable. View return policy

Product Code. 15615640

Brand: Honeywell-Riedel-de Haen 472328100ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -15°C
Density 0.827g/mL at 25°C
Boiling Point 194°C
Flash Point 80°C (176°F)
Odor Characteristic
Packaging Glass Bottle
Linear Formula CH3(CH2)7OH
Refractive Index n20/D 1.429
Quantity 100 mL
Beilstein 1697461
Vapor Density 4.5 (vs air)
Formula Weight 130.23g/mol
Percent Purity ≥99%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material 1-Octanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.