Learn More
1-Octanethiol, Sigma-Aldrich™
Product Code. 15814641
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814641 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15814641 Supplier Sigma-Aldrich Supplier No. 8218610100
 Restricted Item
Estimated Shipment: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-88-6
Grade Research
Molecular Formula CH3(CH2)7SH
Molecular Weight (g/mol) 146.29
Specifications
Melting Point -49°C
Density 0.84 g/cm3 at 20°C
Boiling Point 199°C/1013 hPa
MDL Number MFCD00004912
Quantity 100 mL
Synonym 1-Octanethiol, n-Octyl mercaptan, 1-Octanethiol