missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Naphthyl PP1 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | 1-Naphthyl PP1 hydrochloride |
| Physical Form | Solid |
| CAS | 956025-47-1 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H20ClN5 |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | 1-NA-PP 1 hydrochloride |
| Solubility Information | DMSO : ≥ 73 mg/mL (206.30 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction