Learn More
Melford 1-Naphthyl Phosphate
Product Code. 30018585
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018585 5 g 5g
30018584 25 g 25g
30018583 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30018585 Supplier Melford Supplier No. N420405.0
 Restricted Item
Estimated Shipment: 28-10-2026
Substrate for determaination of phosphatase.
Substrate for determaination of phosphatase. Protect from moisture.
Specifications
Quantity 5 g