Learn More
1-Methylimidazole-2-carboxaldehyde, 98%
Product Code. 15421368
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421368 1 g 1g
15441368 250 mg 250mg
15431368 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15421368 Supplier Thermo Scientific Alfa Aesar Supplier No. H55201.03
Request Bulk or Custom Format
CAS: 13750-81-7 | C5H6N2O | 110.12 g/mol
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water.
Notes
Air sensitive. Store in a cool, dry place. Store in a tightly closed container. Store under nitrogen.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13750-81-7
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD01321308
InChI Key UEBFLTZXUXZPJO-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-2-carbaldehyde, 1-methyl-2-imidazolecarboxaldehyde, 1-methylimidazole-2-carboxaldehyde, 1h-imidazole-2-carboxaldehyde, 1-methyl, 2-formyl-1-methylimidazole, imidazole-2-carboxaldehyde, 1-methyl, pubchem7614, zlchem 1231, 2-formyl-l-methylimidazole, acmc-209ce6
PubChem CID 139575
IUPAC Name 1-methylimidazole-2-carbaldehyde
SMILES CN1C=CN=C1C=O
Specifications
Melting Point 36°C to 39°C
Boiling Point 70°C to 74°C (1 mmHg)
Flash Point 102°C (216°F)
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 110.12
Percent Purity 98%
Chemical Name or Material 1-Methylimidazole-2-carboxaldehyde