Learn More
1-Methyl-4-nitro-1H-imidazole-5-carboxamide, TRC
Product Code. 30470520
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470520 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30470520 Supplier Toronto Research Chemicals Supplier No. TRCM323745500MG
 Restricted Item
Estimated Shipment: 08-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Methyl-4-nitro-1H-imidazole-5-carboxamide
CAS 5413-88-7
Molecular Formula C5 H6 N4 O3
Quantity 500 mg
InChI Formula InChI=1S/C5H6N4O3/c1-8-2-7-5(9(11)12)3(8)4(6)10/h2H,1H3,(H2,6,10)
SMILES Cn1cnc(c1C(=O)N)[N+](=O)[O-]
IUPAC Name 3-methyl-5-nitroimidazole-4-carboxamide
Molecular Weight (g/mol) 170.13
Formula Weight 170.044