Learn More
1-Methyl-2-(tri-n-butylstannyl)pyrrole, 98%, Thermo Scientific Chemicals
Product Code. p-7033990
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15488087
Brand: Thermo Scientific Alfa Aesar H51438.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 118486-97-8
Molecular Formula C17H33NSn
Molecular Weight (g/mol) 370.168
MDL Number MFCD01319022
InChI Key DINAKCGOEKXDTP-UHFFFAOYSA-N
Synonym 1-methyl-2-tributylstannyl pyrrole, 1-methyl-2-tributylstannyl-1h-pyrrole, 1-methyl-2-tributylstannylpyrrole, 1h-pyrrole,1-methyl-2-tributylstannyl, 1-methylpyrrol-2-yl tributyltin, 1-methyl-2-tri-n-butylstannyl pyrrole, n-methyl-2-tributylstannyl-1h-pyrrole, acmc-20aotm, tributyl-1-methylpyrrol-2-yl stannane, tributylstannyl-1-methylpyrrole
PubChem CID 2769397
IUPAC Name tributyl-(1-methylpyrrol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CN1C
Specifications
Density 1.122
Flash Point 98°C (208°F)
Refractive Index 1.511
Quantity 250 mg
UN Number UN2788
Solubility Information Not miscible or difficult to mix.
Formula Weight 370.15
Percent Purity 98%
Chemical Name or Material 1-Methyl-2-(tri-n-butylstannyl)pyrrole