Learn More
1-Methoxy-2-propanol, 99+%
Product Code. 11304308
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 4 L
Unit Size:
1 L
16 L
250mL
4 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304308 1 L 1 L
11394298 250 mL 250mL
11353316 250 mL 250mL
11363316 1 L 1 L
11314308 4 L 4 L
11324308 4 x 4 L 16 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11304308 Supplier Thermo Scientific Alfa Aesar Supplier No. 041547.K2
 Restricted Item
In Stock
Add to basket
CAS: 107-98-2 | C4H10O2 | 90.12 g/mol
1-Methoxy-2-propanol is used as a reagent in the synthesis of 2-amino-3-carboxy-4-phenylthiophenes, which acts as a protein kinase C inhibitors. It is also used as a reagent in the synthesis of metolachlor. It acts as a good biological indicator. Further, it is used as a solvent and an antifreeze agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107-98-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004537
InChI Key ARXJGSRGQADJSQ-UHFFFAOYNA-N
Synonym 1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
PubChem CID 7900
IUPAC Name 1-methoxypropan-2-ol
SMILES COCC(C)O
Specifications
Melting Point °C to 100°C
Density 0.926 g/mL
Boiling Point 119°C to 120°C
Flash Point 35°C (95°F)
Assay Percent Range >99%
Refractive Index 1.403
Quantity 1 L
UN Number UN3092
Beilstein 1731270
Solubility Information Miscible with water,methanol and ether.
Formula Weight 90.12
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-propanol