missing translation for 'onlineSavingsMsg'
Learn More

1-Iodoheptane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog Number p-7027449
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
4282-40-0
C7H15I
226.101
MFCD00001104
LMHCYRULPLGEEZ-UHFFFAOYSA-N
heptyl iodide, n-heptyl iodide, heptane, 1-iodo, iodoheptane, n-heptyliodide, iodo heptane, 1-iodanylheptane, 1-iodomeptane, 1-iodo heptane, 1-iodo-heptane
20274
1-iodoheptane
CCCCCCCI
This item is not returnable. View return policy
4282-40-0
C7H15I
226.101
MFCD00001104
LMHCYRULPLGEEZ-UHFFFAOYSA-N
heptyl iodide, n-heptyl iodide, heptane, 1-iodo, iodoheptane, n-heptyliodide, iodo heptane, 1-iodanylheptane, 1-iodomeptane, 1-iodo heptane, 1-iodo-heptane
20274
1-iodoheptane
CCCCCCCI

1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -48°C
Density 1.379
Boiling Point 203°C to 205°C
Flash Point 78°C (174°F)
Refractive Index 1.49
Quantity 50 g
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 226.1
Percent Purity 98%
Chemical Name or Material 1-Iodoheptane, stabilized with copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'