Learn More
1-Iodo-4-nitrobenzene, TRC
Product Code. 30393741
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393741 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30393741 Supplier Toronto Research Chemicals Supplier No. TRCI71563310G
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Iodo-4-nitrobenzene
CAS 636-98-6
Molecular Formula C6H4INO2
Quantity 10 g
InChI Formula InChI=1S/C6H4INO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H
SMILES [O-][N+](=O)c1ccc(I)cc1
IUPAC Name 1-iodo-4-nitrobenzene
Molecular Weight (g/mol) 249.01
Formula Weight 248.9287